missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPI Polyclonal Antibody

LIPI Polyclonal Antibody
Brand: Invitrogen PA5101651
This item is not returnable.
View return policy
Description
LIPI Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| LIPI | |
| Polyclonal | |
| Unconjugated | |
| LIPI | |
| cancer/testis antigen 17; CT17; D930038D03Rik; lipase I; lipase member I; lipase, member I; LIPI; LPD lipase; lpd1; LPDL; Membrane-associated phosphatidic acid-selective phospholipase A1-beta; mPA-PLA1 beta; PLA1C; PRED5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 149998 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6XZB0 | |
| LIPI | |
| A synthesized peptide derived from human LIPI(Accession Q6XZB0), corresponding to amino acid residues D288-Y338. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction