missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIPF Polyclonal Antibody
GREENER_CHOICE

Product Code. 15545902
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545902 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15545902 Supplier Invitrogen™ Supplier No. PA543148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ISQMITYWGY PNEEYEVVTE DGYILEVNRI PYGKKNSGNT GQRPVVFLQH Sequence homology: Dog: 77%; Guinea Pig: 91%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 93%; Rat: 86%.

NELF (nasal embryonic luteinizing hormone-releasing hormone factor) is a 530 amino acid transcription factor involved in the migration of LHRH neurons, outgrowth of olfactory axons and suppression of transcription elongation. NELF is found in the peripheral and central nervous system during embryonic development, and is highly expressed in adult testis, kidney and brain. Known to couple NMDA receptor signaling to the nucleus, NELF knockdown impaired GnRH neuronal migration of NLT cells in vitro and the gene encoding NELF has been linked to the development of Idiopathic hypogonadotropic hypogonadism (IHH), a disorder resulting in impaired pubertal maturation and reproductive function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPF
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIPF
Gene Accession No. P07098
Gene Alias 2310051B21Rik; AV082900; DGL; EDL; EL; endothelial lipase; gastric lipase; gastric triacylglycerol lipase; GL; HGL; HLAL; lingual lipase; lipase F; lipase F, gastric type; lipase, gastric; LIPE; LIPF; MGC138477; MGC142271; prelingual lipase; RNLIP; Rnlp; triacylglycerol lipase
Gene Symbols LIPF
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LIPF (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8513
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.