missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21133-1-AP-20UL
This item is not returnable.
View return policy
Description
The Lipase family belongs to one of the most robust genetic superfamilies found in living organisms that includes esterases and thioesterases. Lipase gene products are related by tertiary structure rather than primary amino acid sequence. Balancing the composition and the transport of lipoproteins in human plasma is essential for normal body function and is mediated in part by Hepatic Lipase, also known as HL or LIPC. Rare deficiencies in Hepatic Lipase have been identified in humans which lead to pathologic levels of circulating lipoprotein particles; this condition is associated with coronary artery disease (CAD). Hepatic Lipase is regulated by thyroid hormones and has a dual function as a triglyceride hydrolase and a ligand/bridging factor for receptor-mediated lipoprotein uptake. Hepatic Lipase localizes to the endothelial surfaces of extrahepatic tissues.Specifications
| LIPC | |
| Polyclonal | |
| Unconjugated | |
| LIPC | |
| HDLCQ12, Hepatic lipase, Hepatic triacylglycerol lipase, HL, HTGL, Lipase member C, lipase, hepatic, LIPC, LIPH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15450, 24538, 3990 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07867, P11150, P27656 | |
| LIPC | |
| LIPC Fusion Protein Ag15538 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction