missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIPA Monoclonal Antibody (9G7F12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515278
This item is not returnable.
View return policy
Description
MA5-15278 targets LAL in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15278 immunogen is purified recombinant fragment of LAL expressed in E. Coli.
This gene encodes lipase A, the lysosomal acid lipase (also known as cholesterol ester hydrolase). This enzyme functions in the lysosome to catalyze the hydrolysis of cholesteryl esters and triglycerides. Mutations in this gene can result in Wolman disease and cholesteryl ester storage disease. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| LIPA | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| P38571 | |
| LIPA | |
| Purified recombinant fragment of LAL expressed in E. Coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| 9G7F12, 7G6D7 | |
| Unconjugated | |
| LIPA | |
| AA960673; Acid cholesteryl ester hydrolase; CESD; Chole; Chole2; cholesterol ester hydrolase; Cholesterol esterase (pancreatic) see D3Wox12 D3Wox13 D3Wox26 and D3Mgh25; cholesteryl esterase; HGCLAS; HUSSY-01; LAL; LAS; LIAS; Lip1; Lip-1; Lipa; Lipase A; lipase A, lysosomal acid type; lipase A, lysosomal acid, cholesterol esterase; lipoate synthase; Lipoic acid synthase; lipoic acid synthetase; lipoyl synthase, mitochondrial; lip-syn; LS; lysosomal acid lipase; lysosomal acid lipase 1; lysosomal acid lipase A; lysosomal acid lipase/cholesteryl ester hydrolase; pancreatic cholesterol esterase; PDHLD; RP11-341B24.1; Sterol esterase | |
| Mouse | |
| Protein G | |
| RUO | |
| 3988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction