missing translation for 'onlineSavingsMsg'
Learn More

Liothyronine Sodium, Mikromol™

Product Code. 17993266
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993266 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17993266 Supplier Mikromol Supplier No. MM0626.00
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Liothyronine Sodium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Liothyronine sodium
CAS 55-06-1
Analyte or Component Names Liothyronine sodium
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H11 I3 N O4 . Na
Certifications/Compliance ISO 17034
Synonym Liothyronine Sodium, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt (1:1), 3-[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-L-alanine monosodium salt, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine monosodium salt), 3,3',5-Triiodo-L-thyronine sodium salt, 3,3',5-Triiodothyronine sodium salt, 3,5,3'-Triiodo-L-thyronine monosodium salt, Basoprocin, Cynomel, Cyomel, Cytobin, Cytomel, Cytomel sodium, Cytomine, Ibiothyron, L-Triiodothyronine sodium salt, Liothyroniinde sodium salt, Liothyronin sodium, Liothyronine sodium, Sodium 3,3',5-triiodo-L-thyronine, Sodium 3,5,3'-triiodothyronine, Sodium L-3,3',5-triiodothyronine, Sodium L-triiodothyronine, Sodium T3, Sodium liothyronine, Sodium triiodo-L-thyronine, Sodium triiodothyronine, T3 Sodium salt, Tertroxin, Triiodothyronine sodium salt, Triostat, Triothyrone, Levothyroxine Sodium Impurity A, Levothyroxine Impurity A
InChI Formula InChI=1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1
SMILES [Na+].N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(=O)[O-]
IUPAC Name sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
Molecular Weight (g/mol) 672.96
Quantity 50 mg
Formula Weight 672.772 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.