missing translation for 'onlineSavingsMsg'
Learn More

Liothyronine sodium, MedChemExpress

Product Code. 16471976
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471976 500 mg 500mg
16468016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471976 Supplier Medchem Express Supplier No. HYA0070
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Liothyronine sodium is an active form of thyroid hormone. Liothyronine sodium is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.

Liothyronine sodium is an active form of thyroid hormone. Liothyronine sodium is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.

Specifications

Chemical Name or Material Liothyronine sodium
CAS 55-06-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H11I3NNaO4
Quantity 500 mg
Synonym Triiodothyronine sodium3,3',5-Triiodo-L-thyronine sodiumT3 sodium
Solubility Information 0.1 M NaOH : 12.5 mg/mL (18.57 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 5.56 mg/mL (8.26 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES N[C@@H](CC1=CC(I)=C(C(I)=C1)OC2=CC=C(O)C(I)=C2)C(O[Na])=O
Molecular Weight (g/mol) 672.96
Formula Weight 672.96
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.