missing translation for 'onlineSavingsMsg'
Learn More

Liothyronine, MedChemExpress

Product Code. p-200097112
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16478016

Brand: Medchem Express HYA0070A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Liothyronine is an active form of thyroid hormone. Liothyronine is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.

Liothyronine is an active form of thyroid hormone. Liothyronine is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.

Specifications

Chemical Name or Material Liothyronine
CAS 6893-02-3
Color White
Purity Grade Notes Research
Molecular Formula C15H12I3NO4
Quantity 100 mg
Synonym Triiodothyronine 3,3',5-Triiodo-L-thyronine T3
Solubility Information DMSO : 50 mg/mL (76.81 mM; Need ultrasonic) ∣0.1 M NaOH : 16.67 mg/mL (25.61 mM; ultrasonic and warming and adjust pH to 11 with NaOH and heat to 60°C)
SMILES OC1=C(I)C=C(OC2=C(I)C=C(C[C@H](N)C(O)=O)C=C2I)C=C1
Molecular Weight (g/mol) 650.97
Formula Weight 650.97
Percent Purity 98.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.