missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liothyronine Isopropyl Ester, Mikromol™
Description
Liothyronine Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Liothyronine Isopropyl Ester |
| Analyte or Component Names | Liothyronine Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 I3 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Liothyronine Isopropyl Ester |
| InChI Formula | InChI=1S/C18H18I3NO4/c1-9(2)25-18(24)15(22)7-10-5-13(20)17(14(21)6-10)26-11-3-4-16(23)12(19)8-11/h3-6,8-9,15,23H,7,22H2,1-2H3/t15-/m0/s1 |
| SMILES | CC(C)OC(=O)[C@@H](N)Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1 |
| IUPAC Name | propan-2-yl (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate |
| Molecular Weight (g/mol) | 693.05 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?