missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Lionheart FX Automated Microscope
GREENER_CHOICE

Product Code. 17719814 Shop All Agilent BioTek Products
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17719814

Brand: Agilent BioTek LFXWSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. For Research Use Only. Not for use in diagnostic procedures.

Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. It offers up to 60x air; 60x and 100x oil immersion magnification, with fluorescence, brightfield, color brightfield, and phase contrast channels for maximum application reach

An optional environment control cover provides incubation to 40°C and effective containment for CO2/O2 control. A humidity chamber optimizes conditions for long-term live cell imaging applications, and an available dual reagent injector facilitates rapid kinetic assays.

TRUSTED_SUSTAINABILITY

Specifications

Type Cell Imager
Camera CCD
For Use With (Application) Live Cell Imaging
Model LFX
Light Source High Power LED
Software Gen5 Microplate Reader and Imager
Depth (English) 18.3 in.
Depth (Metric) 46.5 cm
Height (English) 14 in.
Height (Metric) 35.6 cm
Width (English) 17.9 in.
Width (Metric) 45.5 cm
Certifications/Compliance CE and TUV marked. RoHS compliant.
Warranty 1 Year
Frequency 50/60 Hz
Voltage 110/220 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.