Learn More
Agilent BioTek Lionheart FX Automated Microscope
Shop All Agilent BioTek Products

Beskrivning
Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. It offers up to 60x air; 60x and 100x oil immersion magnification, with fluorescence, brightfield, color brightfield, and phase contrast channels for maximum application reach
An optional environment control cover provides incubation to 40 °C and effective containment for CO2/O2 control. A humidity chamber optimizes conditions for long-term live cell imaging applications, and an available dual reagent injector facilitates rapid kinetic assays.
Certifikat
CE and TUV
Uppfyllande
RoHS
Specifikationer
Specifikationer
| Type | Cell Imager |
| Camera | 16-bit Gray Scale, Sony CCD, 1.25 Megapixel |
| For Use With (Application) | Live Cell Imaging |
| Model | LFX |
| Light Source | High Power LED |
| Software | Gen5 Microplate Reader and Imager |
| Depth (English) | 18.3 in. |
| Depth (Metric) | 46.5 cm |
| Height (English) | 14 in. |
| Height (Metric) | 35.6 cm |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.