missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lintopride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
20 mg
Unit Size:
10mg
20mg
5mg
Description
Lintopride is a 5HT4 antagonist with moderate 5HT3 antagonist properties.
Specifications
Specifications
| Chemical Name or Material | Lintopride |
| CAS | 107429-63-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C14H19ClN4O2 |
| Quantity | 5 mg |
| SMILES | O=C(NCC1=NCCN1CC)C2=CC(Cl)=C(N)C=C2OC |
| Molecular Weight (g/mol) | 310.78 |
| Formula Weight | 310.78 |
| Percent Purity | 96.38% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction