missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Linopirdine dihydrochloride

Product Code. 30001292
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001292 10 mg 10mg
18025382 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001292 Supplier R&D Systems™ Supplier No. 1999/10
  • ONLINE EXCLUSIVE PRICE
    £216.95
    £194.25 / 10mg
    Save £22.70
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. KV7 (KCNQ) channel blocker

Blocker of KV7 (KCNQ) voltage-gated potassium channels; blocks KV7.1 + 7.3 (KCNQ2 + 3)/ M-currents (IC50 = 4 - 7 μM) and KV7.1 (KCNQ1) homomeric channels (IC50 = 8.9 μM). Augments hippocampal ACh release and is a cognitive enhancer following oral administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113168-57-3
Molecular Formula C26H23Cl2N3O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 464.39
Purity >99%
Molecular Weight (g/mol) 464.39
Chemical Name or Material Linopirdine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.