missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linolenic acid, 99%

Product Code. 10489052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10489052 1 g Glass bottle 1g
10279353 500 mg Glass bottle 500mg
10787192 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10489052 Supplier Thermo Scientific Chemicals Supplier No. 302820010
  • ONLINE EXCLUSIVE PRICE
    £97.30
    £87.65 / 1g
    Save £9.65
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 463-40-1 | C18H30O2 | 278.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 463-40-1
Molecular Formula C18H30O2
Molecular Weight (g/mol) 278.44
MDL Number MFCD00065720
InChI Key DTOSIQBPPRVQHS-PDBXOOCHSA-N
Synonym linolenic acid, alpha-linolenic acid, linolenate, 9z,12z,15z-octadeca-9,12,15-trienoic acid, a-linolenic acid, cis,cis,cis-9,12,15-octadecatrienoic acid, all-cis-9,12,15-octadecatrienoic acid, alpha-linolenate, 9-cis,12-cis,15-cis-octadecatrienoic acid, z,z,z-9,12,15-octadecatrienoic acid
PubChem CID 5280934
ChEBI CHEBI:27432
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoic acid
SMILES CCC=CCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -11.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 230.0°C to 232.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2CH=CH)3(CH2)7CO2H
Refractive Index 1.48
Quantity 1 g
Beilstein 02, IV, 1781
Merck Index 15, 5561
Specific Gravity 0.91
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 278.44
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Linolenic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Linolenic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.