missing translation for 'onlineSavingsMsg'
Learn More

Linoleic Acid, TRC

Product Code. 30004270
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004270 10 g 10g
30011422 1 g 1g
17919544 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004270 Supplier Toronto Research Chemicals Supplier No. TRCL46749510G
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18 H32 O2
Molecular Weight (g/mol) 280.45
Synonym 9,12-Octadecadienoic acid (9Z,12Z)- (9CI, ACI), (9Z,12Z)-9,12-Octadecadienoic acid (ACI), 9,12-Octadecadienoic acid (Z,Z)- (ZCI), Linoleic acid (8CI), (9Z,12Z)-Octadec-9,12-dienoic acid, (Z,Z)-9,12-Octadecadienoic acid, 9,12-Octadecadienoic acid, (Z,Z)-, 9,12-Octadecanoic acid, 9-cis,12-cis-Linoleic acid, 9Z,12Z-Linoleic acid, 9Z,12Z-Octadecadienoic acid, all-cis-9,12-Octadecadienoic acid, C18:2, CA 1726, cis,cis-Linoleic acid, cis-9,cis-12-Octadecadienoic acid, cis-δ9,12-Octadecadienoic acid, Emersol 315, Extra Linoleic 90, LAD 180-6, Linolic acid, MeSH ID: D019787, Polylin 515, Ronacare ASC 3, Unifac 6550, α-Linoleic acid
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-
Formula Weight 280.2402
Chemical Name or Material Linoleic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.