missing translation for 'onlineSavingsMsg'
Learn More

Linoleic acid, tech. 75%

Product Code. 11472318
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472318 250 g 250g
11452318 10 g 10g
11462318 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472318 Supplier Thermo Scientific Alfa Aesar Supplier No. A15265.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 60-33-3 | C18H32O2 | 280.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Density 0.902
Boiling Point 230°C (16 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 75%
Odor Weak, Characteristic
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Refractive Index 1.4699
Quantity 250 g
Beilstein 1727101
Sensitivity Air sensitive
Merck Index 14,5505
Formula Weight 280.45
Percent Purity 75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Linoleic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.