missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linoleic acid, 70%

Product Code. 10157293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157293 2.5 kg Plastic drum 2.5kg
10147750 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157293 Supplier Thermo Scientific Chemicals Supplier No. 413190025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.45
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Acidity 197 to 203mg KOH/g
Melting Point -5.0°C
Color Yellow
Density 0.9000g/mL
Boiling Point 229.0°C to 230.0°C (16.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 2% max. >C18 (GC)
Packaging Plastic drum
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Refractive Index 1.4687 to 1.4707
Quantity 2.5 kg
Beilstein 02, IV, 1754
Merck Index 15, 5560
Specific Gravity 0.9
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether
Formula Weight 280.45
Percent Purity 60%
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material Linoleic acid, tech., 60%
Show More Show Less
compliance-icons
Product Identifier
  • Linoleic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.