missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linoleic Acid Sodium Salt, MP BIOMEDICALS
Shop All MP Biomedicals Products
Click to view available options
:
Linoleic Acid Sodium Salt
Description
- Linoleic acid (LA) is an unsaturated n-6 fatty acid; it belongs to one of the two families of essential fatty acids.
- The body cannot synthesize linoleic acid from other food components.
Application Notes:
- Linoleic acid can be used to show the antioxidant effect of natural phenols.
Usage Statement:
- Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
- Antioxidant
Specifications
Specifications
| Quantity | 5 g |
| Molecular Weight (g/mol) | 302.4 g/mol |
| Percent Purity | 99.00% |
| Physical Form | Powder or Waxy Solid |
Safety and Handling
Product Identifier
- Linoleic Acid Sodium Salt
- Warning
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : Linoleic Acid SodiumSalt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction