missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Linking Clips for Micro Test Tube Rack

Product Code. 12047002
Click to view available options
Color:
Light Gray
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 12047002

Brand: Kartell™ 0029500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Kartell™ Linking Clips for Micro Test Tube Rack is provided in packs of 6 and allows racks to be attached in a row or back to back.

  • Enable attachment in rows or back to back to each other rack
  • Excellent for preparatin work
  • Autoclavable

Specifications

Color Light Gray
Material Polypropylene
For Use With (Equipment) Kartell™ 296
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.