missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Linistat Linear Stainer, 220Vac - 240Vac

Product Code. 12993745 Shop All Epredia Products
This item is not returnable. View return policy
Product Code. 12993745 Supplier Epredia™ Supplier No. B1000201
  • ONLINE EXCLUSIVE PRICE
    £5950.00
    £4461.21 / Pack of 1
    Save £1488.79
    25% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Linistat is a compact and efficient linear stainer that is ideally suited to H&E frozen sections. It is designed to fit on top of most cryostats to streamline workflow during frozen sectioning.

  • Efficient footprint fits on top of most cryostats for streamlining workflow during frozen sectioning
  • Features include 14 staining stations (20 seconds each), running water and a 13 slide capacity collection tank
  • Staining time is under five minutes
  • For situations that require hand staining, the operator may dip through the stations manually, eliminating the need to set up separate staining dishes
  • Color-coded slide clips correspond to the colored slides and colored frozen section embedding media, Epredia™ Cryochrome™
TRUSTED_SUSTAINABILITY

Specifications

Slide Capacity 13
No. of Reagent Stations 14
For Use With (Equipment) Cryostat
Weight (English) 16 lb.
Weight (Metric) 7.3 kg
Height (English) 11 in.
Height (Metric) 27 cm
Length (English) 25.6 in.
Length (Metric) 64 cm
Width (English) 7 in.
Width (Metric) 16 cm
Frequency 50/60 Hz
Voltage 220/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.