missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linifanib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Linifanib (ABT-869) is a potent and orally active multi-target inhibitor of VEGFR and PDGFR family with IC50s of 4, 3, 66, and 4 nM for KDR, FLT1, PDGFRβ, and FLT3, respectively. Linifanib shows prominent antitumor activity. Linifanib has much less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases. Linifanib is a specific miR-10b inhibitor that blocks miR-10b biogenesis.
Specifications
Specifications
| Chemical Name or Material | Linifanib |
| CAS | 796967-16-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H18FN5O |
| Quantity | 200 mg |
| Synonym | ABT-869 AL-39324 |
| Solubility Information | DMSO : 13.16 mg/mL (35.06 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC=C(C2=C3C(N)=NNC3=CC=C2)C=C1)NC4=C(C=CC(C)=C4)F |
| Molecular Weight (g/mol) | 375.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction