missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Linifanib
GREENER_CHOICE

Product Code. 18799036
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18799036 50 mg 50mg
18758316 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18799036 Supplier Tocris Bioscience™ Supplier No. 7743/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent inhibitor of PDGFRβ, KDR, FLT3 and CSF-1R

Linifanib is a potent receptor tyrosine kinase (RTK) inhibitor. It inhibits PDGFRβ, KDR, FLT3, CSF-1R (IC50 values are 2, 4, 4 and 7 nM, respectively). Linifanib inhibits angiogenesis in vascularized micro-organs and potently inhibits VEGF-stimulated endothelial cell proliferation (IC50 = 0.2 nM). Linifanib also promotes the generation and reprogramming of iPSCs from somatic cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-(3-Amino-1 H-indazol-4-yl)phenyl]-N'-(2-fluoro-5-methylphenyl)urea
CAS 796967-16-3
Quantity 50 mg
Target PDGF R beta
Molecular Formula C21H18FN5O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.