missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ LINGO3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565368
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: LWIVQRRKTLN FDGRLPACAT PAEVRGDALR NLPDSVLFEY FVCRKPKIRE RRLQRVTATA GEDVRFLCR Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
LINGO3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include extracellular matrix; extracellular space; integral component of membrane.
Tekniske data
| LINGO3 | |
| Polyclonal | |
| Unconjugated | |
| LINGO3 | |
| hCG2040376; LERN2; leucine rich repeat and Ig domain containing 3; leucine rich repeat neuronal 6B; leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 3; Leucine-rich repeat neuronal protein 2; leucine-rich repeat neuronal protein 6B; LINGO3; LRRN6B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 645191 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0C6S8 | |
| LINGO3 | |
| Recombinant Human LINGO3. Recombinant protein control fragment (Product #RP-100012). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion