missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LINGO-2 Antibody (382007), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36791P
This item is not returnable.
View return policy
Description
LINGO-2 Monoclonal antibody specifically detects LINGO-2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| LINGO-2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 158038 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 382007 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LRRN6Cleucine rich repeat neuronal 6C | |
| Mouse myeloma cell line NS0-derived recombinant human LINGO-2, Cys28-Leu542, Accession # Q7L985 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction