missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Linezolid

Product Code. 18732644
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732644 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732644 Supplier Tocris Bioscience™ Supplier No. 3765/10
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibiotic; inhibits protein synthesis in gram-positive bacteria

Oxazolidinone antibiotic. Inhibits bacterial protein synthesis prior to chain initiation. Displays potent antibacterial activity against a variety of multidrug-resistant gram-positive microbes in vitro and in vivo. Identified as targeting human host cell proteins that interact with SARS-CoV-2.
TRUSTED_SUSTAINABILITY

Specifications

CAS 165800-03-3
Quantity 10 mg
Molecular Formula C16H20FN3O4
Formula Weight 337.35
Purity 0.98
Molecular Weight (g/mol) 337.351
Chemical Name or Material Linezolid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.