missing translation for 'onlineSavingsMsg'
Learn More

Linezolid, MedChemExpress

Product Code. 16462906
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462906 25 mg 25mg
16434946 100 mg 100mg
16430277 50 mg 50mg
16413918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16462906 Supplier Medchem Express Supplier No. HY1039425mg
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Linezolid (PNU-100766) is the first member of the class of oxazolidinone synthetic antibiotic. Linezolid acts by inhibiting the initiation of bacterial protein synthesis. Linezolid is used for the treatment of serious infections caused by Gram-positive bacteria that are resistant to several other antibiotics.

Linezolid (PNU-100766) is the first member of the class of oxazolidinone synthetic antibiotic. Linezolid acts by inhibiting the initiation of bacterial protein synthesis. Linezolid is used for the treatment of serious infections caused by Gram-positive bacteria that are resistant to several other antibiotics.

Specifications

Chemical Name or Material Linezolid
Physical Form Solid
CAS 165800-03-3
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H20FN3O4
For Use With (Application) COVID-19-immunoregulation
Synonym PNU-100766
Solubility Information DMSO : ≥ 100 mg/mL (296.43 mM)
SMILES O=C(O[C@H]1CNC(C)=O)N(C1)C2=CC(F)=C(N3CCOCC3)C=C2
Molecular Weight (g/mol) 337.35
Formula Weight 337.35
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.