missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade

Product Code. 15454705
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454705 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15454705 Supplier Thermo Scientific Chemicals Supplier No. J67830.XF
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear, colorless
Content And Storage -30°C to -10°C
Product Type Linear polyacrylamide solution
Quantity 1 mL
Synonym LPA solution for DNA/RNA precipitation, Molecular Biology Grade;
Form Liquid
At what temperature should I store Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade?

Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade should be stored at -20 degrees Celsius.

What are the instructions for use for Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade?

We recommend that you use the following instructions:
1. Add 1-2 µL of Linear polyacrylamide to 500 µL DNA solution in more than 0.1 M salt (acetate). 1-2 µL (5-10 µg) of 5 mg/mL Linear polyacrylamide (LPA) solution is adequate for 500 µL DNA or RNA solution.
2. Add 2.5 times the sample volume of ethanol. Vortex the mixture or mix gently and incubate in a freezer for 10-15 minutes.
3. Centrifuge for 10 minutes at top speed in a microcentrifuge. A visible pellet will form.
4. Carefully decant or remove the supernatant. The linear polyacrylamide pellet does not stick tightly on the bottom of the microcentrifuge tube. Be careful not to discard the pellet when you remove the supernatant.
5. Carefully wash the pellet with ethanol and let dry.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.