Learn More
Linear alkylbenzenesulfonic acid, 97%
Description
Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 3 x 1 kg |
| UN Number | UN2586 |
| Solubility Information | It is soluble in water. 300,000mg/L at 20°C . |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Chemical Name or Material | Linear alkylbenzenesulfonic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.