missing translation for 'onlineSavingsMsg'
Learn More

Linear alkylbenzenesulfonic acid, 97%

Product Code. 11337318
Click to view available options
Quantity:
250 g
1 kg
3 x 1 kg
Unit Size:
1kg
250g
3kg
This item is not returnable. View return policy

Product Code. 11337318

Brand: Thermo Scientific Alfa Aesar 030574.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68584-22-5 | C18H30O3S | 326.495 g/mol

Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68584-22-5
Molecular Formula C18H30O3S
Molecular Weight (g/mol) 326.495
MDL Number MFCD00147445
InChI Key QJRVOJKLQNSNDB-UHFFFAOYSA-N
Synonym p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid
PubChem CID 29249
IUPAC Name 4-dodecan-3-ylbenzenesulfonic acid
SMILES CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Quantity 250 g
UN Number UN2586
Solubility Information It is soluble in water. 300,000mg/L at 20°C .
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linear alkylbenzenesulfonic acid
compliance-icons compliance-icons
Product Identifier
  • Linear alkylbenzenesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Linear alkylbenzenesulfonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.