missing translation for 'onlineSavingsMsg'
Learn More

Linderane, TRC

Product Code. 30180300
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180300 10 mg 10mg
30179895 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180300 Supplier Toronto Research Chemicals Supplier No. TRCL46634010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13476-25-0
Molecular Formula C15 H16 O4
Molecular Weight (g/mol) 260.29
Synonym 2H-10,1a-(Epoxymethano)oxireno[4,5]cyclodeca[1,2-b]furan-12-one, 3,6,10,10a-tetrahydro-5,9-dimethyl-, (1aS,4E,10S,10aS)-, 2H-10,1a-(Epoxymethano)oxireno[4,5]cyclodeca[1,2-b]furan-12-one, 3,6,10,10a-tetrahydro-5,9-dimethyl-, [1aS-(1aR*,4E,10R*,10aR*)]-, 4βH-Germacra-1(10),7,11-trien-15-oic acid, 4,5β:8,12-diepoxy-6α-hydroxy-, γ-lactone, (E)- (8CI), Linderane (7CI), (1aS,4E,10S,10aS)-3,6,10,10a-Tetrahydro-5,9-dimethyl-2H-10,1a-(epoxymethano)oxireno[4,5]cyclodeca[1,2-b]furan-12-one
SMILES C\C\1=C/CC[C@]23O[C@H]2[C@@H](OC3=O)c4c(C)coc4C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H16O4/c1-8-4-3-5-15-13(19-15)12(18-14(15)16)11-9(2)7-17-10(11)6-8/h4,7,12-13H,3,5-6H2,1-2H3/b8-4+/t12-,13-,15-/m0/s1
Formula Weight 260.1049
Chemical Name or Material Linderane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.