missing translation for 'onlineSavingsMsg'
Learn More

Linalyl acetate, 95%, synthetic

Product Code. 10400131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10400131 500 mL Glass bottle 500mL
10378080 5 mL Glass bottle 5mL
10490121 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10400131 Supplier Thermo Scientific Acros Supplier No. 232305000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-95-7 | C12H20O2 | 196.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-95-7
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.28
MDL Number MFCD00008907
InChI Key UWKAYLJWKGQEPM-UHFFFAOYSA-N
Synonym linalyl acetate, linalool acetate, bergamiol, bergamol, linalol acetate, lynalyl acetate, licareol acetate, bergamot mint oil, acetic acid linalool ester, 3,7-dimethyl-1,6-octadien-3-ol acetate
PubChem CID 8294
ChEBI CHEBI:78333
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl acetate
SMILES CC(=CCCC(C)(C=C)OC(=O)C)C

Specifications

Color Colorless
Density 0.9010g/mL
Boiling Point 220°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2=CHCH2CH2C(CH3)(OOCCH3)CH=CH2
Refractive Index 1.4490 to 1.452
Quantity 500 mL
Merck Index 15, 5551
Specific Gravity 0.901
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,alcohol,ether,diethyl phthal,mineral oil,fixed oils
Formula Weight 196.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Linalyl acetate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Linalyl acetate, synthetic
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.