missing translation for 'onlineSavingsMsg'
Learn More

Linalool, 97%

Product Code. p-153772 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10387050

Brand: Thermo Scientific Acros 125151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-70-6 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-70-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00008906
InChI Key CDOSHBSSFJOMGT-UHFFFAOYSA-N
Synonym linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
PubChem CID 6549
ChEBI CHEBI:17580
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C

Specifications

Color Colorless to Yellow
Density 0.8610g/mL
Boiling Point 199.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Refractive Index 1.4600 to 1.4640
Quantity 100 mL
Beilstein 01, 462
Merck Index 15, 5550
Specific Gravity 0.861
Specific Rotation -2° to +2° (20°C, 589nm) (neat)
Solubility Information Solubility in water: 1.45g/L (25°C). Other solubilities: miscible with alcohol and ether
Viscosity 4.4 mPa.s (25°C)
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linalool, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Linalool
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.