missing translation for 'onlineSavingsMsg'
Learn More

Linaclotide, MedChemExpress

Product Code. 16457408
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16457408

Brand: Medchem Express HY17584

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Linaclotide is a potent and selective guanylate cyclase C agonist; developed for the treatment of constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation.

Linaclotide is a potent and selective guanylate cyclase C agonist

Specifications

Chemical Name or Material Linaclotide
CAS 851199-59-2
Color White
Purity Grade Notes Research
Molecular Formula C59H79N15O21S6
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (32.75 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (10.92 mM; ultrasonic and adjust pH to 2 with HCl)
SMILES O=C([C@@H](NC([C@@](CSSC[C@@H]1N)([H])NC([C@@](CSSC[C@H](N2)C(N[C@H](C(O)=O)CC3=CC=C(O)C=C3)=O)([H])NC([C@@H](NC([C@@H](N4)CCC(O)=O)=O)CC5=CC=C(O)C=C5)=O)=O)=O)CC(N)=O)N6[C@@](C(N[C@H](C(N[C@](CSSC[C@](C4=O)([H])NC1=O)([H])C(NC(C(NCC2=O)=O)[C@H](O)C)=O)=O)C)=O)([H])CCC6
Molecular Weight (g/mol) 1526.74
Formula Weight 1526.74
Percent Purity 97.42%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.