missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIN9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109917
This item is not returnable.
View return policy
Description
Immunogen sequence: TLGGFPVEFL IQVTRLSKIL MIKKEHIKKL REMNTEAEKL KSYSMPISIE FQRRYATIVL ELEQLNKDLN KVLHKVQQYC YELAPDQGLQ PADQPTDMRR RCEEEAQEIV RHANSSTGQP C.
LIN9 belongs to the lin-9 family and acts as a tumor suppressor. It inhibits DNA synthesis. Its ability to inhibit oncogenic transformation is mediated through its association with RB1. LIN9 plays a role in the expression of genes required for the G1/S transition.
Specifications
| LIN9 | |
| Polyclonal | |
| Unconjugated | |
| LIN9 | |
| 2700022J23Rik; BARA; BARPsv; Beta subunit-associated regulator of apoptosis; HuLin-9; LIN9; Lin-9; lin-9 DREAM MuvB core complex component; lin-9 homolog; lin-9 homolog (C. elegans); mLin-9; pRB-associated protein; protein lin-9 homolog; rb related pathway actor; TGS; TGS1; TGS2; TUDOR gene similar 1 protein; TUDOR gene similar protein; Type I interferon receptor beta chain-associated protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 286826, 72568 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5TKA1, Q8C735 | |
| LIN9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-470 of human LIN9 (NP_775106.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction