missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIN9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543640
This item is not returnable.
View return policy
Description
Peptide sequence: SLQKTPVWKG RNTSSAVEMP FRNSKRSRLF SDEDDRQINT RSPKRNQRVA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
LIN9 belongs to the lin-9 family and acts as a tumor suppressor. It inhibits DNA synthesis. Its ability to inhibit oncogenic transformation is mediated through its association with RB1. LIN9 plays a role in the expression of genes required for the G1/S transition.
Specifications
| LIN9 | |
| Polyclonal | |
| Unconjugated | |
| LIN9 | |
| 2700022J23Rik; BARA; BARPsv; Beta subunit-associated regulator of apoptosis; HuLin-9; LIN9; Lin-9; lin-9 DREAM MuvB core complex component; lin-9 homolog; lin-9 homolog (C. elegans); mLin-9; pRB-associated protein; protein lin-9 homolog; rb related pathway actor; TGS; TGS1; TGS2; TUDOR gene similar 1 protein; TUDOR gene similar protein; Type I interferon receptor beta chain-associated protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 286826 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5TKA1 | |
| LIN9 | |
| Synthetic peptide directed towards the N-terminal of human LIN9 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction