missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14656-1-AP-20UL
This item is not returnable.
View return policy
Description
Protein assembly at the post synaptic density (PSD) of neuronal synapses is mediated in part by protein interactions with PDZ (PSD-95/discs large/zona occludens-1) motifs. The Mammalian LIN-7 (MALS) proteins are a family of small synaptic proteins that are the mammalian homologs of LIN-7, a C. elegans protein essential for vulval development. MALS proteins are also known as Veli (Vertebrate Lin-7) proteins. MALS-3 mRNA is most abundant in kidney, followed by brain, liver, thymus and heart. MALS-3 is undetectable in spleen. In cultured cortical neurons, MALS proteins are clustered together with PSD-95 and NMDA-type glutamate receptors. MALS proteins are suggested to have roles in regulating the recruitment of neurotransmitter receptors to the PSD and in the assembly of proteins involved in synaptic vesicle exocytosis and synaptic junctions.Specifications
| LIN7C | |
| Polyclonal | |
| Unconjugated | |
| LIN7C | |
| LIN 7 C, lin 7 homolog C (C. elegans), LIN 7C, LIN7C, MALS 3, MALS3, Mammalian lin seven protein 3, Protein lin 7 homolog C, Veli 3, VELI3, Vertebrate lin 7 homolog 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22343, 55327, 60442 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88952, Q792I0, Q9NUP9 | |
| Lin7c | |
| LIN7C Fusion Protein Ag6265 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction