missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIN7A Monoclonal Antibody (P879)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518305
This item is not returnable.
View return policy
Description
Recommended positive controls: MCF-7, MDA-MB-231. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plays a role in establishing and maintaining the asymmetric distribution of channels and receptors at the plasma membrane of polarized cells. Forms membrane-associated multiprotein complexes that may regulate delivery and recycling of proteins to the correct membrane domains. The tripartite complex composed of LIN7 (LIN7A, LIN7B or LIN7C), CASK and APBA1 may have the potential to couple synaptic vesicle exocytosis to cell adhesion in brain. Ensures the proper localization of GRIN2B (subunit 2B of the NMDA receptor) to neuronal postsynaptic density and may function in localizing synaptic vesicles at synapses where it is recruited by beta-catenin and cadherin. Required to localize Kir2 channels, GABA transporter (SLC6A12) and EGFR/ERBB1, ERBB2, ERBB3 and ERBB4 to the basolateral membrane of epithelial cells.
Specifications
| LIN7A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O14910 | |
| LIN7A | |
| Recombinant protein encompassing a sequence within the center region of human LIN7A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| P879 | |
| Unconjugated | |
| LIN7A | |
| AI848705; hLin-7; lin 7 homolog a; LIN7; lin-7 homolog A (C. elegans); lin-7 homolog A, crumbs cell polarity complex component; Lin7a; lin-7A; lin-7-Ba; Mals1; MALS-1; mammalian LIN-7 1; mammalian lin-seven protein 1; mLin-7; protein lin-7 homolog A; Tax interaction protein 33; TIP-33; Veli; Veli1; veli-1; veli-1 protein; vertebrate homolog of C. elegans Lin-7 type 1; vertebrate LIN7 homolog 1; vertebrate lin-7 homolog 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8825 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction