missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99481CL1
This item is not returnable.
View return policy
Description
LIN7 Polyclonal antibody specifically detects LIN7 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| LIN7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| hLin-7, LIN7, lin-7 homolog A (C. elegans), LIN-7A, MALS1, MALS-1MGC148143, Mammalian lin-seven protein 1, Tax interaction protein 33, TIP-33mammalian LIN-7 1, veli-1, VELI1protein lin-7 homolog A, vertebrate LIN7 homolog 1, Vertebrate lin-7 homolog 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LIN7. | |
| 0.1 mL | |
| Signal Transduction | |
| 8825 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction