missing translation for 'onlineSavingsMsg'
Learn More

LIN7 Antibody, Biotin, Novus Biologicals™

Product Code. 18367606 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367606 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367606 Supplier Novus Biologicals Supplier No. NBP299481B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIN7 Polyclonal specifically detects LIN7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN7
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias hLin-7, LIN7, lin-7 homolog A (C. elegans), LIN-7A, MALS1, MALS-1MGC148143, Mammalian lin-seven protein 1, Tax interaction protein 33, TIP-33mammalian LIN-7 1, veli-1, VELI1protein lin-7 homolog A, vertebrate LIN7 homolog 1, Vertebrate lin-7 homolog 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human LIN7.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8825
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.