missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LIN7 Polyclonal specifically detects LIN7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | LIN7 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | hLin-7, LIN7, lin-7 homolog A (C. elegans), LIN-7A, MALS1, MALS-1MGC148143, Mammalian lin-seven protein 1, Tax interaction protein 33, TIP-33mammalian LIN-7 1, veli-1, VELI1protein lin-7 homolog A, vertebrate LIN7 homolog 1, Vertebrate lin-7 homolog 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human LIN7. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?