missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lin28B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16178-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to LIN28B.
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.Specifications
| Lin28B | |
| Polyclonal | |
| Unconjugated | |
| LIN28B | |
| CSDD2, lin 28 homolog B (C. elegans), Lin 28B, Lin28, LIN28B, Protein lin 28 homolog B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 380669, 389421 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q45KJ6, Q6ZN17 | |
| Lin28b | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction