missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIN28B Monoclonal Antibody (A2-4-5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532901
This item is not returnable.
View return policy
Description
LIN28B Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Specifications
| LIN28B | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6ZN17 | |
| LIN28B | |
| Recombinant protein within Human Lin28B aa 73-250. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| A2-4-5 | |
| Unconjugated | |
| LIN28B | |
| 2810403D23Rik; CSDD2; D030047M17Rik; lin-28 homolog B; lin-28 homolog B (C. elegans); Lin-28.2; Lin28b; Lin-28B; Protein lin-28 homolog B | |
| Mouse | |
| Protein A | |
| RUO | |
| 389421 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction