missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN28A Monoclonal Antibody (14E6-4E6), HRP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1016HRP
This item is not returnable.
View return policy
Description
MA1-016-HRP has been successfully used in Western blot applications on human and mouse samples.
LIN28A/B are expressed in various cancers and ES cells. LIN28 is a cytoplasmic protein, that acts as a ′translational enhancer′, driving specific mRNAs to polysomes and thus increasing the efficiency of protein synthesis. LIN28 is a negative regulator of microRNA (miRNA) biogenesis by specifically binding the precursor of let-7 (pre-let-7). Let-7 miRNAs have been implicated in repression of oncogenes such as ras and myc. LIN28 expression decreases during differentiation of ES cells or upon induction of neuronal differentiation by retinoic acid.
Specifications
| LIN28A | |
| Monoclonal | |
| 1 mg/mL | |
| LIN28A | |
| Lin-28 homolog A (C. elegans), RNA-binding protein LIN-28, LIN28A, CSDD1, Zinc finger CCHC domain-containing protein 1, ZCCHC1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79727, 83557 | |
| 4° C, do not freeze | |
| Liquid |
| Western Blot | |
| 14E6-4E6 | |
| HRP | |
| Q9H9Z2, Q8K3Y3 | |
| LIN28A | |
| Full-length human recombit protein expressed in bacteria | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction