missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LIN28 1632, >98%

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Lin28 1632 is an RNA binding protein Lin28 inhibitor (IC50 = 8 μM). Lin28 1632 blocks binding of Lin28 to let-7 pre-micro RNA. Lin28 1632 inhibits stemness and induces differentiation of murine ESCs. Lin28 1632 increases let-7 miRNA and suppresses PD-L1 expression in multiple human cancer cell lines. Lin28 1632 inhibits growth of tumor cell lines and tumor-sphere formation in vitro. Lin28 1632 also inhibits growth of tumor xenografts in mice by enhancing T cell-mediated antitumor immunity . Lin28 1632 is also bromodomain inhibitor. Lin28 1632 inhibits tumor growth and metastasis in vivo in immunodeficient orthotopic mouse models of human breast cancer cell MDA-MB-231.
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-[3-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide |
| CAS | 108825-65-6 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C15H15N5O |
| Formula Weight | 281.31 |
| Purity | >98% |
| Molecular Weight (g/mol) | 281.319 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido