missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LIN-41/TRIM71 Polyclonal antibody specifically detects LIN-41/TRIM71 in Human samples. It is validated for Western Blot,Immunocytochemistry
Specifications
Specifications
| Antigen | LIN-41/TRIM71 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | LIN41, LIN-41, tripartite motif containing 71, tripartite motif-containing protein 71 |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human LIN-41, Val386-Phe868, Accession # Q2Q1W2 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?