missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIMS2 Monoclonal Antibody (OTI5H8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527458
This item is not returnable.
View return policy
Description
LIMS2 Monoclonal Antibody for Western Blot
Adapter protein in a cytoplasmic complex linking beta- integrins to the actin cytoskeleton, bridges the complex to cell surface receptor tyrosine kinases and growth factor receptors. Plays a role in modulating cell spreading and migration.
Specifications
| LIMS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z4I7, Q91XD2 | |
| Lims2 | |
| Human recombinant protein fragment corresponding to amino acids 1-363 of LIMS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H8 | |
| Unconjugated | |
| Lims2 | |
| ILK-binding protein; LGMD2W; LIM and senescent cell antigen like domains 2; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM zinc finger domain containing 2; LIM-like protein 2; Lims2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; Pinch2; PINCH-2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 225341, 55679 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction