missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIMS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550440
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total LIMS1 protein.
The protein encoded by this gene is an adaptor protein which contains five LIM domains, or double zinc fingers. The protein is likely involved in integrin signaling through its LIM domain-mediated interaction with integrin-linked kinase, found in focal adhesion plaques. It is also thought to act as a bridge linking integrin-linked kinase to NCK adaptor protein 2, which is involved in growth factor receptor kinase signaling pathways. Its localization to the periphery of spreading cells also suggests that this protein may play a role in integrin-mediated cell adhesion or spreading. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| LIMS1 | |
| Polyclonal | |
| Unconjugated | |
| LIMS1 | |
| 2310016J22Rik; 4921524A02Rik; AI507642; AU021743; AW551584; C430041B13Rik; LIM and senescent cell antigen-like domains 1; LIM and senescent cell antigen-like-containing domain protein 1; LIM zinc finger domain containing 1; LIMS1; Lims1l; LIM-type zinc finger domains 1; Particularly interesting new Cys-His protein 1; PINCH; PINCH1; PINCH-1; renal carcinoma antigen NY-REN-48; RGD1560732; senescent cell antigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110829, 3987 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P48059, Q99JW4 | |
| LIMS1 | |
| Fusion protein corresponding to a region derived from internal residues of human LIM and senescent cell antigen-like domains 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction