missing translation for 'onlineSavingsMsg'
Learn More

LIMPII/SR-B2 Antibody, FITC, Novus Biologicals™

Product Code. 18417260 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417260 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417260 Supplier Novus Biologicals Supplier No. NBP177326F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIMPII/SR-B2 Polyclonal antibody specifically detects LIMPII/SR-B2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMPII/SR-B2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the center of human LIMP2. The immunogen is located within amino acids 70 - 120 of LIMP2. Amino Acid Squence: EEVGPYTYRELRNKAN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cholesterol Metabolism, Lipid and Metabolism, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 950
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.