missing translation for 'onlineSavingsMsg'
Learn More

LIMPII/SR-B2 Antibody (017), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18320901 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320901 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320901 Supplier Novus Biologicals Supplier No. NBP290381AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LIMPII/SR-B2 Monoclonal specifically detects LIMPII/SR-B2 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMPII/SR-B2
Applications ELISA
Classification Monoclonal
Clone 17
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LIMPII/SR-B2 (Met 1-Thr 432; NP_031670.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cholesterol Metabolism, GPCR, Lipid and Metabolism, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 950
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.