missing translation for 'onlineSavingsMsg'
Learn More

(±)-Limonene, TRC

Product Code. 30138715
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138715 5 g 5g
30136056 25 g 25g
17953845 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138715 Supplier Toronto Research Chemicals Supplier No. TRCL4617535G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-86-3
Molecular Formula C10 H16
Molecular Weight (g/mol) 136.23
Synonym Cyclohexene, 1-methyl-4-(1-methylethenyl)-, p-Mentha-1,8-diene (8CI), 1-Methyl-4-(1-methylethenyl)cyclohexene, (±)-Dipentene, (±)-Limonene, (±)-α-Limonene, 1,8-p-Menthadiene, 1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, 1-Methyl-4-isopropenyl-1-cyclohexene, 1-Methyl-4-isopropenylcyclohexene, 1-Methyl-p-isopropenyl-1-cyclohexene, 4-Isopropenyl-1-methyl-1-cyclohexene, 4-Isopropenyl-1-methylcyclohexene, Cajeputen, Cajeputene, Cinen, Cinene, DL-Limonene, Dipenten, Dipentene, Eulimen, Flavor orange, Florasolv DPE, Goldflush II, Kautschin, Lemosol, Limonen, Limonene, NSC 21446, NSC 844, Nesol, Orange X, Orange flavor, PC 560, Roti-Histol, SF 001, dl-Limonene, α-Limonene
IUPAC Name 1-methyl-4-prop-1-en-2-ylcyclohexene
SMILES CC(=C)C1CCC(=CC1)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3
Formula Weight 136.1252
Chemical Name or Material Limonene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.