missing translation for 'onlineSavingsMsg'
Learn More

(+)-Limonene, 96%, unstabilized

Product Code. p-153575 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
5mL
This item is not returnable. View return policy

Product Code. 10216140

Brand: Thermo Scientific Acros 179390050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5989-27-5 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5989-27-5, 10191-41-0
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
Synonym d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
PubChem CID 440917
ChEBI CHEBI:15382
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -74.0°C
Color Colorless to Yellow
Density 0.8405g/mL
Boiling Point 175.4°C to 176.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4700 to 1.4740
Quantity 5 mL
Beilstein 05, 133
Merck Index 14, 5493
Specific Gravity 0.8405
Specific Rotation Condition +106.00 (20.00°C c=1,CH3OH)
Specific Rotation + 106.00
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol
IUPAC Name (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene
Viscosity 0.846 mPa.s (25°C)
Formula Weight 136.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (+)-Limonene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+)-Limonene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : D-Limonene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.